Order Entry
Germany
ContactUsLinkComponent
L(+)-Glutamic acid
L(+)-Glutamic acid
Catalog # SERA23000.01
Supplier:  Serva
CAS Number:  
undefined
L(+)-Glutamic acid
Catalog # SERA23000.01
Supplier:  Serva
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :147.13 g/mol
Merck Index:13,04482
Boiling Pt:325 °C (1013 hPa)
Melting Pt:160 °C
Density:1.538 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-293-7
CAS Number:56-86-0
Formula:HO₂CCH₂CH₂CH(NH₂)CO₂H
MDL Number:MFCD00002634