L(+)-Glutamic acid ≥99%, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:56-86-0
:
Formula:HO₂CCH₂CH₂CH(NH₂)CO₂H
MDL Number:MFCD00002634
:147.13 g/mol
Merck Index:13,04482
Boiling Pt:325 °C (1013 hPa)
Melting Pt:160 °C
Density:1.538 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-293-7
Specification Test Results
- Identity Test:Passes
- Purity:≥99%
- FTIR:Conforms to Standard
- Moisture Content:≤0.15%
- Optical Rotation (c=10, 2N HCl ):+25° to +35°
- Solubility (10% in 2N HCl):Clear, Colorless Solution
- Arsenic:≤1,5 ppm
- Heavy Metals:≤20 ppm (as Pb)