L(+)-Glutamic acid
Supplier: BACHEM BIOCHEMICA
Certificates
About this item
CAS Number:56-86-0
:
Formula:HO₂CCH₂CH₂CH(NH₂)CO₂H
MDL Number:MFCD00002634
:147.13 g/mol
Merck Index:13,04482
Boiling Pt:325 °C (1013 hPa)
Melting Pt:160 °C
Density:1.538 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-293-7