Organic reference standard, 2,3,3',6-Tetrachlorobiphenyl (PCB No. 59) 35 µg/ml in isooctane, CIL
PROOCIL-PCB-59
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:74472-33-6
- Description:2,3,3',6-Tetrachlorobiphenyl (PCB No. 59) 35 µg/ml in isooctane
- Formula:C₁₂H₆Cl₄
- Description:2,3,3',6-Tetrachlorobiphenyl (PCB No. 59)