Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L(+)-Sodium lactate
L(+)-Sodium lactate ≥98%
L(+)-Sodium lactate ≥98%
L(+)-Sodium lactate ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
L(+)-Sodium lactate ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
867-56-1
:
Formula:
C₃H₅NaO₃
MDL Number:
MFCD00066576
:
112.06 g/mol
Melting Pt:
∼ 160 °C
Storage Temperature:
Ambient
EINECS:
212-762-3
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAL14500
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation