Order Entry
Germany
ContactUsLinkComponent
L(+)-Lactic acid ≥80% in aqueous solution, Sigma-Aldrich®
Corrosive
L(+)-Lactic acid ≥80% in aqueous solution, Sigma-Aldrich®
Catalog # SIAL27715-1L-R
Supplier:  Merck
CAS Number:  
Corrosive
L(+)-Lactic acid ≥80% in aqueous solution, Sigma-Aldrich®
Catalog # SIAL27715-1L-R
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :90.08 g/mol
Density:1.21 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
EINECS:201-196-2
 :GHS05
Storage Temperature:Refrigerator
CAS Number:79-33-4
Formula:C₃H₆O₃
MDL Number:MFCD00064266