Organic reference standard, 2,2',3,4,4',6-Hexachlorobiphenyl (PCB No. 139) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-139
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:56030-56-9
- Description:2,2',3,4,4',6-Hexachlorobiphenyl (PCB No. 139) 35 µg/ml in isooctane
- Formula:C₁₂H₄Cl₆
- Description:2,2',3,4,4',6-Hexachlorobiphenyl (PCB No. 139)