Organic reference standard, 2,3',4',5-Tetrachlorobiphenyl (PCB No. 70) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20007000IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:32598-11-1
- Description:2,3',4',5-Tetrachlorobiphenyl (PCB No. 70) 10 µg/ml in isooctane
- Formula:C₁₂H₆Cl₄
- Description:2,3',4',5-Tetrachlorobiphenyl (PCB No. 70)