Organic reference standard, 2,2',3,5'-Tetrachlorobiphenyl (PCB No. 44) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-44-CS
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:41464-39-5
- Description:2,2',3,5'-Tetrachlorobiphenyl (PCB No. 44) 100 µg/ml in isooctane
- Formula:C₁₂H₆Cl₄
- Description:2,2',3,5'-Tetrachlorobiphenyl (PCB No. 44)