ContactUsLinkComponent
Plate rack, ImmunoSet®
Plate rack, ImmunoSet®
Catalog # ENZOADI801930
Supplier:  ENZO LIFE SCIENCES
undefined
Plate rack, ImmunoSet®
Catalog # ENZOADI801930
Supplier:  ENZO LIFE SCIENCES
Specifications
  • No. of wells:
    96
  • Colour:
    Clear
  • Description:
    IMMUNOSET® plate rack
  • Seq:
    02
  • Pk:
    1
Specifications
About this item
This ImmunoSet® plate rack is a convenient set of five 96 well, clear, high-binding, polystyrene plates and 5 plate sealers.
Use of these plates will provide optimal performance. The use of different types of plates will produce different results and may affect the sensitivity of the assay due to the type of plastic used.