Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L(+)-Glutamine
L(+)-Glutamine
L(+)-Glutamine
:
PanReac AppliChem
L(+)-Glutamine
:
PanReac AppliChem
:
56-85-9
:
:
C₅H₁₀N₂O₃
:
MFCD00008044
:
146.15 g/mol
:
13,04484
:
355 °C (1013 hPa)
:
185…186 °C
:
1.5326 g/cm³ (130 °C)
:
Ambient
:
200-292-1