(S)-(-)-2-Chloropropionic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:29617-66-1
:
Formula:H₃CCHClCOOH
MDL Number:MFCD00064205
:108.52 g/mol
Boiling Pt:187 °C (760 mmHg)
Melting Pt:4 °C
Density:1.24 g/cm³
Flash Pt:107 °C
Storage Temperature:Ambient
EINECS:411-150-5
UN:2511
ADR:8,III
:GHS07
:GHS05
Specification Test Results
- Appearance:Clear light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4340 to 1.4360 (20°C, 589 nm)
- GC:≥97.5 %
- Specific optical rotation:-14° to -15° (25°C, 589 nm) (neat)
- Titration with NaOH:≥97.5 %