Order Entry
Germany
ContactUsLinkComponent
Organic reference standard, Bromoform 1000 µg/ml in methanol
Organic reference standard, Bromoform 1000 µg/ml in methanol
  PROONE5607
 :  
Organic reference standard, Bromoform 1000 µg/ml in methanol
  PROONE5607
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,5 mL
  • CAS No.:
    75-25-2
  • Description:
    Bromoform 1,000 µg/ml in methanol
  • Formula:
    CHBr₃
  • Description:
    Bromoform