0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Iron (II) sulphate heptahydrate
Iron(II) sulphate heptahydrate analytical reagent
Iron(II) sulphate heptahydrate analytical reagent
:
Bernd Kraft
Iron(II) sulphate heptahydrate analytical reagent
:
Bernd Kraft
:
7782-63-0
:
:
FeSO₄·7H₂O
:
MFCD00149719
:
278.02 g/mol
:
13,04091
:
64 °C
:
1.9 g/cm³ (20 °C)
:
231-753-5
:
GHS07
:
Ambient