Nickel(II) nitrate hexahydrate ≥97.0% (by complexometric analysis), crystallised, Purum p.a.
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:13478-00-7
:
Formula:Ni(NO₃)₂·6H₂O
MDL Number:MFCD00149805
:290.79 g/mol
Merck Index:12,06596
Boiling Pt: Dec. (on heating)
Melting Pt:56.7 °C
Density:2.050 g/cm³ (20 °C)
EINECS:236-068-5
UN:2725
ADR:5.1,III
:GHS07
:GHS05
:GHS08
:GHS03
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Assay:97.0 - 103.0 %
- Calcium (Ca):Max. 50 ppm
- Cadmium (Cd):Max. 50 ppm
- Cobalt (Co):Max. 500 ppm
- Copper (Cu):Max. 50 ppm
- Iron (Fe):Max. 50 ppm
- Potassium (K):Max. 100 ppm
- Sodium (Na):Max. 100 ppm
- Lead (Pb):Max. 50 ppm
- Zinc (Zn):Max. 50 ppm
- Chloride (Cl):Max. 50 ppm
- Sulfate (SO₄):Max. 50 ppm