Nickel(II) nitrate hexahydrate 98.0-102.0%, AnalaR NORMAPUR® ACS analytical reagent (max. 0.005% Co)
Catalog # 25873.232
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Assay:98.0 - 102.0 %
- Insoluble substances:Max. 50 ppm
- Cl (Chloride):Max. 10 ppm
- SO4 (Sulphate):Max. 20 ppm
- Ca (Calcium):Max. 50 ppm
- Co (Cobalt):Max. 50 ppm
- Cu (Copper):Max. 5 ppm
- Fe (Iron):Max. 10 ppm
- K (Potassium):Max. 100 ppm
- Mg (Magnesium):Max. 100 ppm
- Mn (Manganese):Max. 20 ppm
- Na (Sodium):Max. 50 ppm
- Pb (Lead):Max. 10 ppm
- Zn (Zinc):Max. 10 ppm
- Conforms to ACS:Passes test
Specification Test Results
About this item
:
:290.79 g/mol
Merck Index:12,06596
Boiling Pt:137 °C (1013 hPa)
Melting Pt:56.7 °C
Density:2.05 g/cm³ (20 °C)
EINECS:236-068-5
UN:2725
ADR:5.1,III
:GHS03
:GHS08
:GHS07
:GHS05
:GHS09
CAS Number:13478-00-7
Formula:Ni(NO₃)₂·6H₂O
MDL Number:MFCD00149805