

Iron(III) nitrate nonahydrate 99.0-101.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7782-61-8
:
Formula:Fe(NO₃)₃·9H₂O
MDL Number:MFCD00149708
:404 g/mol
Merck Index:13,04057
Boiling Pt:125 °C (1013 hPa)
Melting Pt:47 °C
Density:1.68 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-899-5
UN:1466
ADR:5.1,III
:GHS03
:GHS07
Specification Test Results
- Assay:99.0 - 101.0 %
- Free acid:Max. 0.05 meq/g
- Insolubility in water:Max. 0.02 %
- Not precipitated by NH4OH (as SO4):Max. 0.1 %
- Cl (Chloride):Max. 5 ppm
- PO4 (Phosphate):Max. 50 ppm
- SO4 (Sulphate):Max. 50 ppm
- Ca (Calcium):Max. 50 ppm
- Cu (Copper):Max. 50 ppm
- K (Potassium):Max. 50 ppm
- Mg (Magnesium):Max. 10 ppm
- Mn (Manganese):Max. 0.02 %
- Na (Sodium):Max. 50 ppm
- Pb (Lead):Max. 10 ppm
- Zn (Zinc):Max. 10 ppm
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 250 ml Plastic bottle for solids DIN 60 (ZPACK13250250) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
SDS_EN_CH_VWRC24175.pdf SDS_EN_DE_VWRC24175.pdf SDS_EN_NL_VWRC24175.pdf SDS_EN_UK_VWRC24175.pdf sds_VWRC24175
SDS
spec_VWRC24175.233 spec_VWRC24175.260 spec_VWRC24175.260 spec_VWRC24175.260 spec_VWRC24175.290
Product specification
More