Iron(III) nitrate nonahydrate analytical reagent
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:7782-61-8
:
Formula:Fe(NO₃)₃·9H₂O
MDL Number:MFCD00149708
:404 g/mol
Merck Index:13,04057
Boiling Pt:125 °C (1013 hPa)
Melting Pt:47 °C
Density:1.68 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-899-5
UN:1466
ADR:5.1,III
:GHS03
:GHS07