Cobalt(II) nitrate hexahydrate ≥99.999% (metals basis), Puratronic®
10694.09
:
- Pack type:Plastic bottle for solids
- Pk:10 g
- Seq:3236
:
:291.03 g/mol
:14,2444
:55…56 °C
:1.87
:Ambient
:233-402-1
:1477
:5.1,III
:GHS03
:GHS08
:GHS07
:10026-22-9
:Co(NO₃)₂·6H₂O
:MFCD00149647