Nickel(II) chloride hexahydrate
Catalog # HONE13613.9050
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pk:50 kg
- Seq:4418
Specifications
About this item
:
:237.69 g/mol
Merck Index:13,06529
Melting Pt:140 °C
Density:1.84 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-743-0
UN:3288
ADR:6.1,III
:GHS06
:GHS08
:GHS09
CAS Number:7791-20-0
Formula:NiCl₂·6H₂O
MDL Number:MFCD00149809