Order Entry
Germany
ContactUsLinkComponent
Iron(II) chloride tetrahydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Corrosive
Irritant
Iron(II) chloride tetrahydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 0125.0250
Supplier:  Avantor
CAS Number:  
Corrosive
Irritant
Iron(II) chloride tetrahydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 0125.0250
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Assay:
    min. 99.0 %
  • Appearance of solution:
    passes test
  • Arsenic (As):
    max. 0.001 %
  • Copper (Cu):
    max. 0.005 %
  • Zinc (Zn):
    max. 0.005 %
  • Insoluble Matter:
    max. 0.01 %
  • Phosphate (PO4):
    max. 0.001 %
  • Sulfate (SO4):
    max. 0.01 %

Specification Test Results

About this item

 :
Melting Pt:677 °C
Density:1.926 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3260
ADR:8,III
 :198.81 g/mol
EINECS:231-843-4
 :GHS05
 :GHS07
CAS Number:13478-10-9
Formula:FeCl₂·4H₂O
MDL Number:MFCD00149709