
Manganese(II) bromide tetrahydrate 98%
ACRO197130500
:
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
- Appearance:Pink Crystalline powder or crystals
- Titration Argentometric:≥97.5 %
:
:286.81 g/mol
:236-591-9
:GHS07
:Ambient
:10031-20-6
:Br₂Mn·4H₂O
:MFCD00149791