Zinc sulphate heptahydrate, Sigma-Aldrich®
Catalog # SIALZ1001-500G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
Merck Index:12,10293
Melting Pt:100 °C
Density:1.97 g/cm³ (20 °C)
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ200
:287.54 g/mol
EINECS:231-793-3
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:7446-20-0
Formula:ZnSO₄·7H₂O
MDL Number:MFCD00149894