Zinc sulphate heptahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7446-20-0
:
Formula:ZnSO₄·7H₂O
MDL Number:MFCD00149894
Merck Index:12,10293
Melting Pt:100 °C
Density:1.97 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:9,III
:M-Clarity™ quality level = MQ100
:287.54 g/mol
EINECS:231-793-3
UN:3077
:GHS05
:GHS07
:GHS09