Zinc sulphate heptahydrate, extra pure
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:7446-20-0
:
Formula:ZnSO₄·7H₂O
MDL Number:MFCD00149894
Merck Index:12,10293
Melting Pt:100 °C
Density:1.97 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-793-3
UN:3077
ADR:9,III
:287.54 g/mol
:GHS05
:GHS07
:GHS09