Order Entry
Germany
ContactUsLinkComponent
 
Toxic
Health
Environment
Cadmium(II) sulphate 8/3-hydrate, EMSURE® ACS for analysis
Catalog # 1.02027.0100
Supplier:  Merck
CAS Number:  
Toxic
Health
Environment
Cadmium(II) sulphate 8/3-hydrate, EMSURE® ACS for analysis
Catalog # 1.02027.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay (complexometric, CdSO₄.8/3 H₂O):
    98.0 - 102.0 %
  • Insoluble matter:
    ≤ 0.005 %
  • pH-value (5 %; water):
    3.0 - 6.0
  • Chloride (Cl):
    ≤ 0.001 %
  • Total nitrogen (N):
    ≤ 0.0005 %
  • Ca (Calcium):
    ≤ 0.005 %
  • Cu (Copper):
    ≤ 0.0005 %
  • Fe (Iron):
    ≤ 0.0005 %
  • K (Potassium):
    ≤ 0.01 %
  • Na (Sodium):
    ≤ 0.005 %
  • Pb (Lead):
    ≤ 0.002 %
  • Zn (Zinc):
    ≤ 0.002 %

Specification Test Results

About this item

 :
 :256.51 g/mol (anhy)
Merck Index:13,01627
Melting Pt:41 °C
Density:3.79 g/cm³ (20 °C)
EINECS:233-331-6
UN:2570
ADR:6.1,III
 :M-Clarity™ quality level = MQ300
 :GHS06
 :GHS08
 :GHS09
CAS Number:7790-84-3
Formula:3CdSO₄·8H₂O
MDL Number:MFCD02253000