


Cadmium(II) sulphate 8/3-hydrate 98.0-102.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7790-84-3
:
Formula:3CdSO₄·8H₂O
MDL Number:MFCD00151655
:256.51 g/mol (anhy)
Merck Index:14,1627
Melting Pt:>40 °C (-H₂O)
Density:3.08
EINECS:233-331-6
UN:2570
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient