Order Entry
Germany
ContactUsLinkComponent
Terbium trinitrate hexahydrate 99.99+% (trace metals basis)
Terbium trinitrate hexahydrate 99.99+% (trace metals basis)
Certificates
undefined
Terbium trinitrate hexahydrate 99.99+% (trace metals basis)

Specification Test Results

  • Appearance:
    White Granular powder
  • Assay:
    ≥99.99 % (TREO base)
  • Total conc. of imp.as REO:
    ≤0.01 %

Specification Test Results

Related Information