Tetraethylammonium chloride hydrate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56-34-8
:
Formula:C₈H₂₀ClN·xH₂O
MDL Number:MFCD00149992
:165.71 g/mol (anhy)
Merck Index:14,9200
Melting Pt:∼ 110 °C
Density:1.0801
EINECS:200-267-5
:GHS07
Storage Temperature:Ambient