Tetraethylammonium chloride hydrate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56-34-8
:
Formula:C₈H₂₀ClN·xH₂O
MDL Number:MFCD00149992
:165.71 g/mol (anhy)
EINECS:200-267-5
:GHS07
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥98.5 % (on dry substance)
- Loss on drying:≤10 %
- Impurity:≤0.5 % Triethylamine hydrochloride