
Aluminium chloride hexahydrate
Catalog # HONE11016.9025
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pk:25 kg
- Seq:4283
Specifications
About this item
:
:241.43 g/mol
Melting Pt:∼100 °C
Density:2.39 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-208-1
UN:3260
ADR:8,III
:GHS07
CAS Number:7784-13-6
Formula:AlCl₃·6H₂O
MDL Number:MFCD00149134