Magnesium sulphate hydrate ≥99.997% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:22189-08-8
:15244-36-7
Formula:MgSO₄·nH₂O
MDL Number:MFCD00011110
:120.37 g/mol (anhy)
Merck Index:14,5691
Density:1.68
Storage Temperature:Ambient
EINECS:231-298-2