Order Entry
Germany
ContactUsLinkComponent
Capping B 1-Methylimidazol : Tetrahydrofuran (18:82) (w/w) , Novabiochem®
Flammable
Corrosive
Health
Irritant
Capping B 1-Methylimidazol : Tetrahydrofuran (18:82) (w/w) , Novabiochem®
Catalog # 8.57012.4000
Supplier:  Merck
CAS Number:  
Flammable
Corrosive
Health
Irritant
Capping B 1-Methylimidazol : Tetrahydrofuran (18:82) (w/w) , Novabiochem®
Catalog # 8.57012.4000
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    4 L
  • Seq:
    5162

Specifications

About this item

 :
ADR:3 (8),II
 :M-Clarity™ quality level = MQ200
UN:2924
 :GHS02
 :GHS05
 :GHS08
 :GHS07