Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Capping Reagent
Capping A 20% (v/v) n-Methylimidazole in ACN, Novabiochem® for DNA/RNA synthesis
Capping A 20% (v/v) n-Methylimidazole in ACN, Novabiochem® for DNA/RNA synthesis
Capping A 20% (v/v) n-Methylimidazole in ACN, Novabiochem® for DNA/RNA synthesis
Supplier:
Merck
Certificates
Expand image
Capping A 20% (v/v) n-Methylimidazole in ACN, Novabiochem® for DNA/RNA synthesis
Supplier:
Merck
About this item
:
ADR:
3 (8),II
:
M-Clarity™ quality level = MQ200
UN:
2924
:
GHS02
:
GHS06
:
GHS05
Related Information
Documentation
SDS
msds_857002
Documentation