
Strontium hydroxide octahydrate
Catalog # HONE13913.9025
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pk:25 kg
- Seq:4283
Specifications
About this item
:
:265.76 g/mol
Melting Pt:375 °C
Density:1.9 g/cm³ (20 °C)
EINECS:242-367-1
UN:3262
ADR:8,III
:GHS05
CAS Number:1311-10-0
Formula:H₂O₂Sr·8H₂O
MDL Number:MFCD00150395