Calcium chloride dihydrate 99-105% ACS
:10035-04-8
:
:CaCl₂·2H₂O
:MFCD00149613
:147.01 g/mol
:13,01660
:1600 °C (1013 hPa)
:176 °C
:1.85 g/cm³ (20 °C)
:233-140-8
:GHS07
:Ambient
- Ammonium:<0.005%
- Assay:99 – 105%
- Barium:≤0.005%
- Heavy Metals (as Pb):≤0.0005%
- Insolubles:≤0.01%
- Iron:≤0.001%
- Magnesium:≤0.005%
- Oxidizers (as NO₃):≤0.003%
- pH (5%, Water) @25 °C:4.5 – 8.5
- Potassium:≤0.01%
- Sodium:≤0.02%
- Strontium:≤0.1%
- Sulphate:≤0.01%