Aluminium potassium sulphate dodecahydrate ≥97%, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7784-24-9
:
Formula:AlK(SO₄)₂·12H₂O
MDL Number:MFCD00149143
:474.39 g/mol
Merck Index:12,00373
Melting Pt:92.5 °C
Density:1.72 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-141-3
REACH:01-2119960162-44
Specification Test Results
- Assay:Min. 97 %
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 5L Plastic bucket UN Z (ZPACK13247005)
Packaging Information
spec_VWRC21098.293 spec_VWRC21098.293 spec_VWRC21098.293 spec_VWRC21098.362 spec_VWRC21098.362
Product specification
More