Order Entry
Germany
ContactUsLinkComponent
Aluminium potassium sulphate dodecahydrate 99.0-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., Fluka™
Aluminium potassium sulphate dodecahydrate 99.0-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., Fluka™
Catalog # 12623-1KG
CAS Number:  
Aluminium potassium sulphate dodecahydrate 99.0-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., Fluka™
Catalog # 12623-1KG
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay (calculated to the dried substance):
    99.0 - 100.5 %
  • Loss on drying:
    43.0 - 46.0 %
  • Ammonium (NH₄):
    Max. 0.2 %
  • Iron (Fe):
    complying
  • Arsenic (As):
    Max. 0.0002 %
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 0.005 %
  • Appearance of the solution:
    complying

Specification Test Results

About this item

 :
 :474.39 g/mol
Merck Index:12,00373
Melting Pt:92.5 °C
Density:1.75 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-141-3
CAS Number:7784-24-9
Formula:AlK(SO₄)₂·12H₂O
MDL Number:MFCD00149143