Disodium hydrogen phosphate heptahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7782-85-6
:
Formula:HNa₂O₄P·7H₂O
MDL Number:MFCD00149180
:268.07 g/mol
Melting Pt:48 °C
Density:1.7 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:231-448-7