Order Entry
Germany
ContactUsLinkComponent
Sodium carbonate monohydrate ≥99.5% ACS, Fluka™
Irritant
Sodium carbonate monohydrate ≥99.5% ACS, Fluka™
Certificates
Irritant
Sodium carbonate monohydrate ≥99.5% ACS, Fluka™

Specification Test Results

  • Assay:
    Min. 99.5 %
  • Loss on drying (150°C):
    13.0 - 15.0 %
  • Water insoluble matter:
    Max. 0.01 %
  • Calcium (Ca):
    Max. 0.03 %
  • Iron (Fe):
    Max. 5 ppm
  • Potassium (K):
    Max. 0.005 %
  • Magnesium (Mg):
    Max. 0.005 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 0.001 %
  • Phosphate (PO₄):
    Max. 5 ppm
  • S-compounds (as SO₄):
    Max. 0.004 %
  • Silicate (as SiO₂):
    Max. 0.005 %

Specification Test Results