Order Entry
Germany
ContactUsLinkComponent
tetra-Chloroauric(III) acid trihydrate ≥49.0% (Au basis) ACS, Fluka™
Corrosive
Irritant
tetra-Chloroauric(III) acid trihydrate ≥49.0% (Au basis) ACS, Fluka™
Certificates
Corrosive
Irritant
tetra-Chloroauric(III) acid trihydrate ≥49.0% (Au basis) ACS, Fluka™

Specification Test Results

  • Appearance of the substance:
    complying
  • Assay of Au:
    Min. 49.0 %
  • Alkali- and other metals (as SO₄):
    Max. 0.2 %
  • In ether insoluble matter:
    Max. 0.1 %

Specification Test Results

Related Information

Frequently Bought Together