Order Entry
Germany
ContactUsLinkComponent
 
 
p-Toluenesulphonic acid monohydrate for synthesis, Sigma-Aldrich®
 :  Merck
 
 
p-Toluenesulphonic acid monohydrate for synthesis, Sigma-Aldrich®
 :  Merck
 

 

  • Solubility:
    670 g/l (20 °C)
  • Melting Point:
    56 °C Elimination of water of crystallisation
  • Molar Mass:
    190.22 g/mol
  • Bulk Density:
    510 kg/m3
  • Boiling Point:
    140 °C (27 hPa)
  • Vapor Pressure:
    (20 °C)
  • Flash Point:
    180 °C
  • Density:
    0.5 g/cm3
  • pH:
    1 (650 g/l, H2O, 20 °C) (anhydrous substance)
  • Ignition Point:
    600 °C
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive material

 

Frequently Bought Together