
3,5-Dihydroxytoluene monohydrate (colorimetric), Sigma-Aldrich®
:6153-39-5
:
:C₇H₈O₂·1H₂O
:MFCD00149092
:142.15 g/mol
:12,06995
:290 °C (1013 hPa) (lit.)
:56…58 °C (lit.)
:1.290 g/cm³ (4 °C)
:M-Clarity™ quality level = MQ200
:159 °C
:207-984-2
:GHS07
:Room Temperature
- Appearance (Colour):White to Light Brown to Light Orange
- Appearance (Form):Powder
- Solubility (Colour):Colorless to Light Brown
- Solubility (Turbidity)
200 mg + 4 ml H2O:Clear - Water
1H NMR Spectrum:10 - 15 - Purity (HPLC):≥99 %
- Recommended Retest Period:3 years