Order Entry
Germany
ContactUsLinkComponent
1-Hexanesulphonic acid sodium salt monohydrate 99%
Irritant
1-Hexanesulphonic acid sodium salt monohydrate 99%
Certificates
undefined
Irritant
1-Hexanesulphonic acid sodium salt monohydrate 99%
Certificates

Specification Test Results

  • Appearance:
    White Powder or crystals or flakes or crystalline solid
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤9.6 % (100°C, 3 h) (vacuum)
  • Solubility:
    (10 % in water) Clear colorless
  • Titration after Ion exchange:
    ≥98.5 % (on dry substance)

Specification Test Results

Related Information