tri-Sodium citrate dihydrate, extra pure
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:6132-04-3
:
Formula:Na₃C₆H₅O₇·2H₂O
MDL Number:MFCD00150031
:294.1 g/mol
Merck Index:13,08675
Melting Pt:150 °C
Density:1.76 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-675-3
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Potassium chloride, EMSURE® for analysis, Supelco®
Supplier: Merck
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Phosphate Buffered Saline (PBS), pH 7,4
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals