Order Entry
Germany
ContactUsLinkComponent
tri-Sodium citrate dihydrate, BAKER, J.T.Baker®
tri-Sodium citrate dihydrate, BAKER, J.T.Baker®
Catalog # 1758.1000
Supplier:  Avantor
CAS Number:  
undefined
tri-Sodium citrate dihydrate, BAKER, J.T.Baker®
Catalog # 1758.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay (anhydrous substance):
    99.0 - 100.5 %
  • Acidity or Alkalinity:
    passes test
  • Appearance of solution:
    passes test
  • Chloride (Cl):
    max. 0.003 %
  • Heavy Metals (as Pb):
    max. 10 ppm
  • Identification:
    passes test
  • Oxalates (C2O4):
    max. 300 ppm
  • Readily Carbonisable Substances:
    passes test
  • Sulfate (SO4):
    max. 0.005 %
  • Water (H2O):
    11.0 - 13.0 %
  • Tartrate:
    passes test

Specification Test Results

About this item

 :
 :294.1 g/mol
Merck Index:13,08675
Melting Pt:150 °C
Density:1.76 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-675-3
CAS Number:6132-04-3
Formula:Na₃C₆H₅O₇·2H₂O
MDL Number:MFCD00150031