
di-Ammonium oxalate monohydrate 99+% ACS
:6009-70-7
:
:(NH₄)₂C₂O₄·H₂O
:MFCD00149694
:142.11 g/mol
:13,00541
:70 °C (1013 hPa)
:70 °C
:~ 1.5 g/cm³ (20 °C)
:Ambient
:214-202-3
:6.1,III
:GHS07
- Appearance:White Crystalline powder
- Residue after ignition:≤0.02 %
- Titration with KMnO4:99.0 to 101.0 %
- Chloride (Cl):≤0.002 %
- Sulfate (SO4):≤0.002 %
- Iron (Fe):≤2 ppm
- Heavy metals (as Pb):≤5 ppm
- Insoluble matter:≤0.005 %