
Manganese(II) acetate tetrahydrate (∼22% Mn)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6156-78-1
:
Formula:(H₃CCOO)₂Mn·4H₂O
MDL Number:MFCD00062552
:245.09 g/mol
Merck Index:14,5724
Melting Pt:80 °C
Density:1.589
EINECS:211-334-3
:GHS07
Storage Temperature:Ambient