Order Entry
Germany
ContactUsLinkComponent
 
Zinc acetate dihydrate ≥98%, GPR RECTAPUR®
Catalog # 29087.298
Supplier:  VWR Chemicals
CAS Number:  
undefined
Zinc acetate dihydrate ≥98%, GPR RECTAPUR®
Catalog # 29087.298
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 98 %
  • Cl (Chloride):
    Max. 50 ppm
  • SO4 (Sulphate):
    Max. 0.02 %
  • Fe (Iron):
    Max. 20 ppm
  • Pb (Lead):
    Max. 100 ppm

Specification Test Results

About this item

 :
 :219.51 g/mol
Merck Index:12,10256
Melting Pt:237 °C
Density:1.718 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-170-2
UN:3077
ADR:9,III
 :GHS07
 :GHS09
CAS Number:5970-45-6
Formula:(H₃CCOO)₂Zn·2H₂O
MDL Number:MFCD00066961