Sodium acetate trihydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6131-90-4
:
Formula:H₃CCOONa·3H₂O
MDL Number:MFCD00071557
:136.08 g/mol
Merck Index:13,08642
Boiling Pt:120…123 °C (1013 hPa)
Melting Pt:58 °C
Density:1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-823-8
Frequently Bought Together